Product Details
- SNP ID
-
rs144333708
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
5
- Location
-
Chr.1:84369961 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATCACAGAGGGTCTTCTGCACAGA[A/G]GTCAAGTATTCACCTTTGTCATAGG
- Phenotype
-
MIM: 191540
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
UOX
PubMed Links
Gene Details
- Gene
- UOX
- Gene Name
- urate oxidase (pseudogene)
There are no transcripts associated with this gene.
View Full Product Details