Product Details

SNP ID
rs144336229
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:108648770 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGAAGGGTGTGGGAGAGTTCTC[C/T]ATCTTGGCCTTCTCAACCTCTGTGA
Phenotype
MIM: 612178
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM102B PubMed Links

Gene Details

Gene
FAM102B
Gene Name
family with sequence similarity 102 member B
There are no transcripts associated with this gene.

Gene
HENMT1
Gene Name
HEN1 methyltransferase homolog 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102592.1 1399 Missense Mutation ATA,ATG I326M NP_001096062.1
NM_144584.2 1399 Missense Mutation ATA,ATG I326M NP_653185.2
XM_005270411.1 1399 Missense Mutation ATA,ATG I334M XP_005270468.1
XM_017000179.1 1399 Missense Mutation ATA,ATG I170M XP_016855668.1

View Full Product Details