Product Details

SNP ID
rs144341849
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:53921654 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCAAGAGCATTTTGTTATCATTA[C/T]GAGGAAAGATTTGAGACTACTGTTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HSPB11 PubMed Links

Gene Details

Gene
HSPB11
Gene Name
heat shock protein family B (small) member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316935.1 716 Silent Mutation TCA,TCG S144S NP_001303864.1
NM_016126.3 716 Silent Mutation TCA,TCG S144S NP_057210.2

View Full Product Details