Product Details

SNP ID
rs144369082
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:151167401 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTCCCCACTATAACAGTTGCTGC[C/T]GCCGGAAGTACAGGTATGGGACGGG
Phenotype
MIM: 608095 MIM: 605834 MIM: 612112 MIM: 600607
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LYSMD1 PubMed Links

Gene Details

Gene
LYSMD1
Gene Name
LysM domain containing 1
There are no transcripts associated with this gene.

Gene
SCNM1
Gene Name
sodium channel modifier 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204856.1 565 Missense Mutation CGC,TGC R94C NP_001191785.1
NM_024041.3 565 Missense Mutation CGC,TGC R129C NP_076946.1
Gene
TMOD4
Gene Name
tropomodulin 4
There are no transcripts associated with this gene.

Gene
TNFAIP8L2
Gene Name
TNF alpha induced protein 8 like 2
There are no transcripts associated with this gene.

Gene
TNFAIP8L2-SCNM1
Gene Name
TNFAIP8L2-SCNM1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204848.1 565 Missense Mutation CGC,TGC R94C NP_001191777.1
Gene
VPS72
Gene Name
vacuolar protein sorting 72 homolog
There are no transcripts associated with this gene.

View Full Product Details