Product Details

SNP ID
rs144371094
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:248406286 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGCCAATGGGGTTATGATCTTC[C/T]TGATCCAAACAGATTTGCGCCTTCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR2T1 PubMed Links

Gene Details

Gene
OR2T1
Gene Name
olfactory receptor family 2 subfamily T member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030904.1 292 Silent Mutation CTG,TTG L98L NP_112166.1

View Full Product Details