Product Details

SNP ID
rs144423087
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:2591973 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAAGAGCTGCTCATGGGTGAGAT[C/T]CAGGCCGGGCAGCTGCTGGTCCTTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM213B PubMed Links

Gene Details

Gene
FAM213B
Gene Name
family with sequence similarity 213 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195736.1 2602 Intron NP_001182665.1
NM_001195737.1 2602 Intron NP_001182666.1
NM_001195738.1 2602 Intron NP_001182667.1
NM_001195740.1 2602 Intron NP_001182669.1
NM_001195741.1 2602 Intron NP_001182670.1
NM_152371.3 2602 Intron NP_689584.2
XM_006710354.3 2602 Intron XP_006710417.1
XM_011540664.1 2602 Intron XP_011538966.1
XM_011540665.1 2602 Intron XP_011538967.1
XM_011540666.1 2602 Intron XP_011538968.1
Gene
LOC100996583
Gene Name
uncharacterized LOC100996583
There are no transcripts associated with this gene.

Gene
MMEL1
Gene Name
membrane metallo-endopeptidase-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033467.3 2602 Missense Mutation AAT,GAT N708D NP_258428.2
XM_011542122.2 2602 Missense Mutation AAT,GAT N451D XP_011540424.1
XM_017002310.1 2602 Missense Mutation AAT,GAT N708D XP_016857799.1
XM_017002311.1 2602 Missense Mutation AAT,GAT N690D XP_016857800.1
XM_017002312.1 2602 Missense Mutation AAT,GAT N682D XP_016857801.1
XM_017002313.1 2602 Intron XP_016857802.1
XM_017002314.1 2602 Missense Mutation AAT,GAT N665D XP_016857803.1
XM_017002315.1 2602 Missense Mutation AAT,GAT N604D XP_016857804.1

View Full Product Details