Product Details

SNP ID
rs144559086
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:11108251 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGATGAGCAGCTCAACTTGCGTTG[C/G]AACATCCAAAGTGTCATCATGAGAG
Phenotype
MIM: 605960 MIM: 601231
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EXOSC10 PubMed Links

Gene Details

Gene
EXOSC10
Gene Name
exosome component 10
There are no transcripts associated with this gene.

Gene
LOC105376736
Gene Name
uncharacterized LOC105376736
There are no transcripts associated with this gene.

Gene
MTOR
Gene Name
mechanistic target of rapamycin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004958.3 7148 Missense Mutation CCA,GCA P2522A NP_004949.1
XM_005263438.2 7148 Missense Mutation CCA,GCA P2522A XP_005263495.1
XM_011541166.2 7148 Intron XP_011539468.1
XM_017000900.1 7148 Missense Mutation CCA,GCA P2295A XP_016856389.1
XM_017000901.1 7148 Missense Mutation CCA,GCA P2106A XP_016856390.1
XM_017000902.1 7148 Intron XP_016856391.1

View Full Product Details