Product Details

SNP ID
rs144585204
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:1295453 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCCACCCCACACTTACTGCGAG[G/T]GCCAGCCCCTGCGGCCCCTGCGTCG
Phenotype
MIM: 601328
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ACAP3 PubMed Links

Gene Details

Gene
ACAP3
Gene Name
ArfGAP with coiled-coil, ankyrin repeat and PH domains 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030649.2 4354 Missense Mutation ACT,CCT T603P NP_085152.2
XM_005244715.2 4354 Missense Mutation ACT,CCT T603P XP_005244772.1
XM_005244717.3 4354 Missense Mutation ACT,CCT T262P XP_005244774.1
XM_011540606.2 4354 Missense Mutation ACT,CCT T613P XP_011538908.1
XM_011540607.1 4354 Missense Mutation ACT,CCT T613P XP_011538909.1
XM_011540608.2 4354 Missense Mutation ACT,CCT T581P XP_011538910.1
XM_011540609.2 4354 Missense Mutation ACT,CCT T613P XP_011538911.1
XM_017000233.1 4354 Missense Mutation ACT,CCT T414P XP_016855722.1
Gene
MIR6726
Gene Name
microRNA 6726
There are no transcripts associated with this gene.

Gene
SCNN1D
Gene Name
sodium channel epithelial 1 delta subunit
There are no transcripts associated with this gene.

View Full Product Details