Product Details
- SNP ID
-
rs144624013
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
7
- Location
-
Chr.1:158480447 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGTTTTCAGCCTCCCTTTTTGTAG[C/T]GCCAACAAAGTCAATCATTACTTCT
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR10R2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3820678] are located under a probe and SNP(s) [rs61741711] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR10R2
- Gene Name
- olfactory receptor family 10 subfamily R member 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001004472.1 |
570 |
Silent Mutation |
AGC,AGT |
S190S |
NP_001004472.1 |
View Full Product Details