Product Details

SNP ID
rs144630443
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:1217739 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGTCGCCTTTCCCCCTCCGAGCTC[C/T]GCGGCCAGCCGCACGAAGTGGCTAT
Phenotype
MIM: 614282 MIM: 603905 MIM: 600315
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SDF4 PubMed Links

Gene Details

Gene
SDF4
Gene Name
stromal cell derived factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016176.3 1307 Intron NP_057260.2
NM_016547.2 1307 Silent Mutation NP_057631.1
XM_011541556.1 1307 Intron XP_011539858.1
Gene
TNFRSF18
Gene Name
TNF receptor superfamily member 18
There are no transcripts associated with this gene.

Gene
TNFRSF4
Gene Name
TNF receptor superfamily member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003327.3 1307 Intron NP_003318.1
XM_011542074.2 1307 Intron XP_011540376.1
XM_011542075.2 1307 Intron XP_011540377.1
XM_011542076.2 1307 Intron XP_011540378.1
XM_011542077.2 1307 Intron XP_011540379.1
XM_017002231.1 1307 Intron XP_016857720.1
XM_017002232.1 1307 Intron XP_016857721.1

View Full Product Details