Product Details
- SNP ID
-
rs144643375
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
10
- Location
-
Chr.1:152303006 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACCTTTTTGCCTTTCAGTGCCCTC[A/G]GATTGATAATGATAAGAACTAGAAC
- Phenotype
-
MIM: 135940
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FLG
PubMed Links
Gene Details
- Gene
- FLG
- Gene Name
- filaggrin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002016.1 |
11916 |
Silent Mutation |
TCC,TCT |
S3960S |
NP_002007.1 |
View Full Product Details