Product Details

SNP ID
rs144661113
Assay Type
Functionally Tested
NCBI dbSNP Submissions
17
Location
Chr.1:248361592 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGGTGAAACCTCATGGACAACAT[C/T]ACCTGGATGGCCAGCCACACTGGAT
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
OR2T4 PubMed Links

Gene Details

Gene
OR2T4
Gene Name
olfactory receptor family 2 subfamily T member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004696.1 Intron NP_001004696.1

View Full Product Details