Product Details

SNP ID
rs144669476
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:185167573 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAATATTAACCTTCTTGAAACAC[A/C]GATTTCATCCCATTTCTCCTCTGCT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SWT1 PubMed Links

Gene Details

Gene
SWT1
Gene Name
SWT1, RNA endoribonuclease homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105518.1 Intron NP_001098988.1
NM_017673.6 Intron NP_060143.4
XM_005245271.1 Intron XP_005245328.1
XM_005245273.1 Intron XP_005245330.1
XM_011509657.1 Intron XP_011507959.1
XM_017001527.1 Intron XP_016857016.1
XM_017001528.1 Intron XP_016857017.1
XM_017001529.1 Intron XP_016857018.1
XM_017001530.1 Intron XP_016857019.1

View Full Product Details