Product Details

SNP ID
rs144732881
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:169793039 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTTACTTCATCAATCACCATACT[A/G]TTATAATCTTGAAAGTGAATTTCTT
Phenotype
MIM: 615255
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf112 PubMed Links

Gene Details

Gene
C1orf112
Gene Name
chromosome 1 open reading frame 112
There are no transcripts associated with this gene.

Gene
METTL18
Gene Name
methyltransferase like 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320199.1 916 Silent Mutation AAC,AAT N219N NP_001307128.1
NM_001320201.1 916 Silent Mutation AAC,AAT N219N NP_001307130.1
NM_033418.3 916 Silent Mutation AAC,AAT N219N NP_219486.1
XM_006711627.3 916 Silent Mutation AAC,AAT N219N XP_006711690.1

View Full Product Details