Product Details

SNP ID
rs144782990
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:70263987 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTAGATTTAAACACAATAATCAAC[A/G]TAACAATTATAGAGTTTCCAGTTAT
Phenotype
MIM: 615125
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD13C PubMed Links

Gene Details

Gene
ANKRD13C
Gene Name
ankyrin repeat domain 13C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030816.4 Intron NP_110443.3
XM_005271234.3 Intron XP_005271291.1
XM_005271235.3 Intron XP_005271292.1
XM_006710929.3 Intron XP_006710992.1
XM_017002413.1 Intron XP_016857902.1
XM_017002414.1 Intron XP_016857903.1
XM_017002415.1 Intron XP_016857904.1
XM_017002416.1 Intron XP_016857905.1

View Full Product Details