Product Details

SNP ID
rs144785768
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:158547488 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATAGCAATGACCATGAGGGCCAA[A/G]AAGAAGTCCACCATCTCAGCCTGTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR6Y1 PubMed Links

Gene Details

Gene
OR6Y1
Gene Name
olfactory receptor family 6 subfamily Y member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005189.1 618 Silent Mutation TTC,TTT F206F NP_001005189.1

View Full Product Details