Product Details
- SNP ID
-
rs144794319
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:27100329 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGTTACTGCCCCTTGGGGAAGAAC[A/G]GTTCTCCCTCCCCAGGCTCAGGGTG
- Phenotype
-
MIM: 107310
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SLC9A1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs4266911] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SLC9A1
- Gene Name
- solute carrier family 9 member A1
View Full Product Details