Product Details

SNP ID
rs144800595
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:35608723 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTCTTTCAAGAGGAGGCAGCCA[C/T]TAGGTGACAACCAAGGGAAGGCTCA
Phenotype
MIM: 602175
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PSMB2 PubMed Links

Gene Details

Gene
PSMB2
Gene Name
proteasome subunit beta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199779.1 Intron NP_001186708.1
NM_001199780.1 Intron NP_001186709.1
NM_002794.4 Intron NP_002785.1

View Full Product Details