Product Details

SNP ID
rs144909438
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:204199195 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCTGCAGGCATCATCTGCTTAC[C/T]GCACCCAGGAAGGGGATGTTGCAGA
Phenotype
MIM: 603286
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GOLT1A PubMed Links

Gene Details

Gene
GOLT1A
Gene Name
golgi transport 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198447.1 546 Silent Mutation GCA,GCG A120A NP_940849.1
XM_017000314.1 546 Silent Mutation GCA,GCG A120A XP_016855803.1
Gene
KISS1
Gene Name
KiSS-1 metastasis-suppressor
There are no transcripts associated with this gene.

View Full Product Details