Product Details

SNP ID
rs144945086
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:152614009 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTGCTGCAGGTCCCACCGATGC[C/T]GGCGCCAGAGCTCCAACTCCTGTGA
Phenotype
MIM: 612613 MIM: 612614
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LCE3A PubMed Links

Gene Details

Gene
LCE3A
Gene Name
late cornified envelope 3A
There are no transcripts associated with this gene.

Gene
LCE3B
Gene Name
late cornified envelope 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178433.1 199 Missense Mutation CGG,TGG R67W NP_848520.1

View Full Product Details