Product Details

SNP ID
rs144956634
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:37566858 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAATAAAAACCTTTTAAACATTA[C/T]TGCTTTTGTCTGAATTTTTTGCGTT
Phenotype
MIM: 602135 MIM: 609365
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNALI1 PubMed Links

Gene Details

Gene
DNALI1
Gene Name
dynein axonemal light intermediate chain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003462.3 2356 Intron NP_003453.2
XM_005271172.2 2356 Intron XP_005271229.1
Gene
GNL2
Gene Name
G protein nucleolar 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323623.1 2356 Silent Mutation CAA,CAG Q799Q NP_001310552.1
NM_001323624.1 2356 Silent Mutation CAA,CAG Q548Q NP_001310553.1
NM_013285.2 2356 Silent Mutation CAA,CAG Q731Q NP_037417.1

View Full Product Details