Product Details

SNP ID
rs145025672
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:153678863 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTTCACACTCGGGTCCTCTCCAG[A/C]CCGACGTTCTCCTGGCACCCACCTG
Phenotype
MIM: 603181 MIM: 108960
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ILF2 PubMed Links

Gene Details

Gene
ILF2
Gene Name
interleukin enhancer binding factor 2
There are no transcripts associated with this gene.

Gene
NPR1
Gene Name
natriuretic peptide receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000906.3 247 UTR 5 NP_000897.3
XM_005245218.1 247 UTR 5 XP_005245275.1
XM_017001374.1 247 UTR 5 XP_016856863.1

View Full Product Details