Product Details

SNP ID
rs145035907
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:229594420 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCCACGAGCTCGCTGGAGGAGCC[A/G]TCGGCAGGTGCACTGCAGTAAGTGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TAF5L PubMed Links

Gene Details

Gene
TAF5L
Gene Name
TATA-box binding protein associated factor 5 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025247.1 1779 Intron NP_001020418.1
NM_014409.3 1779 Silent Mutation GAC,GAT D549D NP_055224.1
XM_005273099.4 1779 Silent Mutation GAC,GAT D549D XP_005273156.1
XM_005273100.2 1779 Silent Mutation GAC,GAT D457D XP_005273157.1
XM_011544162.1 1779 Silent Mutation GAC,GAT D478D XP_011542464.1
XM_011544163.1 1779 Silent Mutation GAC,GAT D457D XP_011542465.1
XM_011544164.2 1779 Silent Mutation GAC,GAT D457D XP_011542466.1
XM_017001045.1 1779 Silent Mutation GAC,GAT D457D XP_016856534.1

View Full Product Details