Product Details
- SNP ID
-
rs145041158
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
15
- Location
-
Chr.1:207759702 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTAAAGGATCAGTAGCAATTTGGAG[C/T]GGTAAGCCCCCAATATGTGAAAGTA
- Phenotype
-
MIM: 120920
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CD46
PubMed Links
Gene Details
- Gene
- CD46
- Gene Name
- CD46 molecule
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002389.4 |
628 |
Silent Mutation |
AGC,AGT |
S151S |
NP_002380.3 |
NM_153826.3 |
628 |
Silent Mutation |
AGC,AGT |
S151S |
NP_722548.1 |
NM_172350.2 |
628 |
Silent Mutation |
AGC,AGT |
S151S |
NP_758860.1 |
NM_172351.2 |
628 |
Silent Mutation |
AGC,AGT |
S151S |
NP_758861.1 |
NM_172352.2 |
628 |
Silent Mutation |
AGC,AGT |
S151S |
NP_758862.1 |
NM_172353.2 |
628 |
Silent Mutation |
AGC,AGT |
S151S |
NP_758863.1 |
NM_172359.2 |
628 |
Silent Mutation |
AGC,AGT |
S151S |
NP_758869.1 |
NM_172361.2 |
628 |
Silent Mutation |
AGC,AGT |
S151S |
NP_758871.1 |
XM_011509563.1 |
628 |
Silent Mutation |
AGC,AGT |
S151S |
XP_011507865.1 |
XM_011509564.1 |
628 |
Silent Mutation |
AGC,AGT |
S151S |
XP_011507866.1 |
XM_017001308.1 |
628 |
Silent Mutation |
AGC,AGT |
S151S |
XP_016856797.1 |
XM_017001309.1 |
628 |
Silent Mutation |
AGC,AGT |
S151S |
XP_016856798.1 |
XM_017001310.1 |
628 |
Silent Mutation |
AGC,AGT |
S151S |
XP_016856799.1 |
View Full Product Details