Product Details

SNP ID
rs145048352
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:28553689 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCATCTCCAGAGCCTTTGCCACCA[C/T]GGGGGAGACCGTAATGAGCGTCAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRNAU1AP PubMed Links

Gene Details

Gene
TRNAU1AP
Gene Name
tRNA selenocysteine 1 associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017846.4 171 Missense Mutation ACG,ATG T26M NP_060316.1
XM_017001542.1 171 Intron XP_016857031.1

View Full Product Details