Product Details

SNP ID
rs145081583
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:47416738 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGCGAGCCGGGCAACCCGGGCAA[A/G]GGCAACTACTGGACGCTGGACCCCG
Phenotype
MIM: 601094
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FOXE3 PubMed Links

Gene Details

Gene
FOXE3
Gene Name
forkhead box E3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012186.2 667 Silent Mutation AAA,AAG K141K NP_036318.1
Gene
LINC01389
Gene Name
long intergenic non-protein coding RNA 1389
There are no transcripts associated with this gene.

View Full Product Details