Product Details

SNP ID
rs145233928
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:111500236 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGGACTTAGCCGTCTTGAACTCC[C/T]GTCCATAAAATGCACCTGTCTCTTT
Phenotype
MIM: 600445
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADORA3 PubMed Links

Gene Details

Gene
ADORA3
Gene Name
adenosine A3 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000677.3 1438 Missense Mutation CAG,CGG Q224R NP_000668.1
NM_001302678.1 1438 UTR 3 NP_001289607.1
NM_001302679.1 1438 Missense Mutation CAG,CGG Q79R NP_001289608.1
Gene
TMIGD3
Gene Name
transmembrane and immunoglobulin domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081976.2 1438 Intron NP_001075445.1
NM_001302680.1 1438 Intron NP_001289609.1
NM_020683.6 1438 Intron NP_065734.5

View Full Product Details