Product Details

SNP ID
rs145257885
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:52423973 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCTCTTGGTGGGTGGGGACAAC[A/G]CTCTCTACACCGACGGGTGGCACAT
Phenotype
MIM: 617031 MIM: 613692
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRPF38A PubMed Links

Gene Details

Gene
PRPF38A
Gene Name
pre-mRNA processing factor 38A
There are no transcripts associated with this gene.

Gene
ZCCHC11
Gene Name
zinc finger CCHC-type containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009881.2 4463 Missense Mutation CGT,TGT R1634C NP_001009881.1
NM_015269.2 4463 Missense Mutation CGT,TGT R1633C NP_056084.1
XM_005270676.2 4463 Intron XP_005270733.1
XM_005270678.2 4463 Intron XP_005270735.1
XM_005270679.2 4463 Intron XP_005270736.1
XM_005270680.2 4463 Missense Mutation CGT,TGT R1629C XP_005270737.1
XM_005270681.2 4463 Intron XP_005270738.1
XM_005270682.2 4463 Intron XP_005270739.1
XM_005270683.2 4463 Intron XP_005270740.1
XM_006710498.2 4463 Intron XP_006710561.1
XM_006710499.3 4463 Intron XP_006710562.1
XM_006710500.1 4463 Intron XP_006710563.1
XM_011541097.1 4463 Intron XP_011539399.1
XM_011541098.1 4463 Intron XP_011539400.1
XM_011541099.1 4463 Intron XP_011539401.1
XM_011541100.2 4463 Intron XP_011539402.1
XM_011541101.2 4463 Intron XP_011539403.1
XM_017000803.1 4463 Intron XP_016856292.1
XM_017000804.1 4463 Missense Mutation CGT,TGT R1602C XP_016856293.1
XM_017000805.1 4463 Missense Mutation CGT,TGT R1591C XP_016856294.1
XM_017000806.1 4463 Intron XP_016856295.1
XM_017000807.1 4463 Missense Mutation CGT,TGT R1558C XP_016856296.1
XM_017000808.1 4463 Intron XP_016856297.1
XM_017000809.1 4463 Missense Mutation CGT,TGT R1523C XP_016856298.1
XM_017000810.1 4463 Missense Mutation CGT,TGT R1518C XP_016856299.1
XM_017000811.1 4463 Missense Mutation CGT,TGT R1512C XP_016856300.1
XM_017000812.1 4463 Missense Mutation CGT,TGT R1511C XP_016856301.1
XM_017000813.1 4463 Missense Mutation CGT,TGT R1507C XP_016856302.1
XM_017000814.1 4463 Missense Mutation CGT,TGT R1401C XP_016856303.1
XM_017000815.1 4463 Intron XP_016856304.1
XM_017000816.1 4463 Intron XP_016856305.1
XM_017000817.1 4463 Intron XP_016856306.1
XM_017000818.1 4463 Intron XP_016856307.1

View Full Product Details