Product Details

SNP ID
rs145267654
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:248406238 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGGTCTTATTTTTGCCATCATC[G/T]CTATCATCTTCTTCACCGCACTGAT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR2T1 PubMed Links

Gene Details

Gene
OR2T1
Gene Name
olfactory receptor family 2 subfamily T member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030904.1 244 Missense Mutation GCT,TCT A82S NP_112166.1

View Full Product Details