Product Details

SNP ID
rs145409270
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:43384518 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGCTTCTTCACCTTGCGCCTTA[C/G]AGGGATAGCCAGAATGAAACCCAGG
Phenotype
MIM: 607956 MIM: 615463
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MED8 PubMed Links

Gene Details

Gene
MED8
Gene Name
mediator complex subunit 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001653.2 886 UTR 3 NP_001001653.1
NM_052877.4 886 Missense Mutation CTA,GTA L280V NP_443109.2
NM_201542.4 886 UTR 3 NP_963836.2
Gene
SZT2
Gene Name
seizure threshold 2 homolog (mouse)
There are no transcripts associated with this gene.

View Full Product Details