Product Details

SNP ID
rs145591989
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:26053932 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGTGGACTCTTCCTCTTCCTGAT[A/C]TTCTTCTTCAATGAATTCTTCCTCT
Phenotype
MIM: 609617 MIM: 606131
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SLC30A2 PubMed Links

Gene Details

Gene
SLC30A2
Gene Name
solute carrier family 30 member 2
There are no transcripts associated with this gene.

Gene
TRIM63
Gene Name
tripartite motif containing 63
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032588.3 1152 Missense Mutation GAT,TAT D338Y NP_115977.2
XM_017002559.1 1152 Missense Mutation GAT,TAT D338Y XP_016858048.1

View Full Product Details