Product Details

SNP ID
rs145594154
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:150997045 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACATACGTGTGACTATTCAGGGA[C/T]CGGGAGTTGAGAACCAGAAACCCAC
Phenotype
MIM: 603319
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANXA9 PubMed Links

Gene Details

Gene
ANXA9
Gene Name
annexin A9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003568.2 2588 Intron NP_003559.2
Gene
FAM63A
Gene Name
family with sequence similarity 63 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040217.2 2588 UTR 3 NP_001035307.1
NM_001163258.1 2588 UTR 3 NP_001156730.1
NM_001163259.1 2588 UTR 3 NP_001156731.1
NM_001163260.1 2588 UTR 3 NP_001156732.1
NM_001319998.1 2588 UTR 3 NP_001306927.1
NM_018379.4 2588 UTR 3 NP_060849.2
XM_005245321.3 2588 UTR 3 XP_005245378.1
XM_005245325.3 2588 UTR 3 XP_005245382.1
XM_005245327.3 2588 UTR 3 XP_005245384.1
XM_005245328.3 2588 UTR 3 XP_005245385.1
XM_011509757.2 2588 UTR 3 XP_011508059.1
XM_011509759.2 2588 UTR 3 XP_011508061.1
XM_017001770.1 2588 UTR 3 XP_016857259.1
XM_017001771.1 2588 UTR 3 XP_016857260.1
XM_017001772.1 2588 UTR 3 XP_016857261.1
XM_017001773.1 2588 UTR 3 XP_016857262.1
XM_017001774.1 2588 UTR 3 XP_016857263.1
XM_017001775.1 2588 UTR 3 XP_016857264.1
XM_017001776.1 2588 UTR 3 XP_016857265.1
XM_017001777.1 2588 UTR 3 XP_016857266.1

View Full Product Details