Product Details

SNP ID
rs145664368
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:2056497 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCCCACCCAGGTGACCCTTGCAC[A/G]GTGTCCTCCCAGATGGAGCTGGAAG
Phenotype
MIM: 176982
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105378591 PubMed Links

Gene Details

Gene
LOC105378591
Gene Name
uncharacterized LOC105378591
There are no transcripts associated with this gene.

Gene
PRKCZ
Gene Name
protein kinase C zeta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033581.1 364 Intron NP_001028753.1
NM_001033582.1 364 Intron NP_001028754.1
NM_001242874.1 364 Intron NP_001229803.1
NM_002744.4 364 Silent Mutation ACA,ACG T69T NP_002735.3
XM_011541773.1 364 Silent Mutation ACA,ACG T69T XP_011540075.1
XM_011541774.1 364 Intron XP_011540076.1
XM_011541775.2 364 Silent Mutation ACA,ACG T69T XP_011540077.1
XM_011541776.1 364 Intron XP_011540078.1
XM_011541778.1 364 Intron XP_011540080.1
XM_017001789.1 364 Silent Mutation ACA,ACG T69T XP_016857278.1
XM_017001790.1 364 Silent Mutation ACA,ACG T69T XP_016857279.1
XM_017001791.1 364 Intron XP_016857280.1
XM_017001792.1 364 Intron XP_016857281.1
XM_017001793.1 364 Intron XP_016857282.1
XM_017001794.1 364 Intron XP_016857283.1
XM_017001795.1 364 Intron XP_016857284.1
XM_017001796.1 364 Intron XP_016857285.1
XM_017001797.1 364 Intron XP_016857286.1
XM_017001798.1 364 Intron XP_016857287.1
XM_017001799.1 364 Intron XP_016857288.1
XM_017001800.1 364 Intron XP_016857289.1
XM_017001801.1 364 Intron XP_016857290.1
XM_017001802.1 364 Intron XP_016857291.1
XM_017001803.1 364 Silent Mutation ACA,ACG T69T XP_016857292.1
XM_017001804.1 364 Intron XP_016857293.1
XM_017001805.1 364 Intron XP_016857294.1

View Full Product Details