Product Details

SNP ID
rs145680047
Assay Type
Functionally Tested
NCBI dbSNP Submissions
1
Location
Chr.1:145936466 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCTCTAGACAGTGCCCCCGGCAC[C/A]CTCTGCTCCCGGCTCACCCAGTCCA
Phenotype
MIM: 605313
Polymorphism
C/A, Transversion Substitution
Allele Nomenclature
Literature Links
LIX1L PubMed Links

Gene Details

Gene
LIX1L
Gene Name
limb and CNS expressed 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153713.2 950 Silent Mutation AGC,AGT S286S NP_714924.1
XM_017000316.1 950 Silent Mutation AGC,AGT S254S XP_016855805.1
Gene
RBM8A
Gene Name
RNA binding motif protein 8A
There are no transcripts associated with this gene.

View Full Product Details