Product Details

SNP ID
rs145697009
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:23779910 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAGGCATCATTATAATGGGAGAG[C/G]ATGATGACTCACACCCCTCTGAGAT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PITHD1 PubMed Links

Gene Details

Gene
PITHD1
Gene Name
PITH domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020362.4 344 Missense Mutation CAT,GAT H97D NP_065095.2
XM_011541804.1 344 Missense Mutation CAT,GAT H4D XP_011540106.1
Gene
TCEB3-AS1
Gene Name
TCEB3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details