Product Details

SNP ID
rs145698278
Assay Type
Functionally Tested
NCBI dbSNP Submissions
9
Location
Chr.1:26832588 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGGGCCACTGTCTGTGAAGCAGC[C/T]GCCCTGGAGAAACAGATCGGTGAGG
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ZDHHC18 PubMed Links

Gene Details

Gene
ZDHHC18
Gene Name
zinc finger DHHC-type containing 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032283.2 572 Silent Mutation GCC,GCT A159A NP_115659.1

View Full Product Details