Product Details

SNP ID
rs145724932
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:89262193 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACATACTCTCATCGTTACAGGCAG[A/G]GGAGAGATGAAACAATTGATGAATA
Phenotype
MIM: 611467
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GBP5 PubMed Links

Gene Details

Gene
GBP5
Gene Name
guanylate binding protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134486.2 Intron NP_001127958.1
NM_052942.3 Intron NP_443174.1

View Full Product Details