Product Details

SNP ID
rs145882179
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:10462682 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTTAGCCAGCAGGCGACGATCCC[A/C]TCAAGTTTGCCTTTTCCTGTGACAT
Phenotype
MIM: 601882
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DFFA PubMed Links

Gene Details

Gene
DFFA
Gene Name
DNA fragmentation factor subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004401.2 1257 Intron NP_004392.1
NM_213566.1 1257 UTR 3 NP_998731.1

View Full Product Details