Product Details

SNP ID
rs145909259
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:209828064 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGGCAAACTTGACGTTTTCGCT[A/C]TGGGCAAACGCGGGAGCCGGAGCCA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DIEXF PubMed Links

Gene Details

Gene
DIEXF
Gene Name
digestive organ expansion factor homolog (zebrafish)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014388.6 98 Missense Mutation ATG,CTG M1L NP_055203.4
XM_006711275.3 98 Missense Mutation ATG,CTG M1L XP_006711338.1
XM_011509418.2 98 UTR 5 XP_011507720.1

View Full Product Details