Product Details

SNP ID
rs145924837
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:103564417 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTATTTAAATTCTGATACTCACC[C/T]GTATCCAGTTAGTTTGTTGTACTCC
Phenotype
MIM: 104660
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACTG1P4 PubMed Links

Gene Details

Gene
ACTG1P4
Gene Name
actin gamma 1 pseudogene 4
There are no transcripts associated with this gene.

Gene
AMY2B
Gene Name
amylase, alpha 2B (pancreatic)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020978.4 Intron NP_066188.1
Gene
RNPC3
Gene Name
RNA binding region (RNP1, RRM) containing 3
There are no transcripts associated with this gene.

View Full Product Details