Product Details

SNP ID
rs145959705
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:149929654 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGGCCTTCCCCTCAGGTAGCAG[C/T]GTCTCCAGAGCCTGATCTGGGGTCC
Phenotype
MIM: 605593
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MTMR11 PubMed Links

Gene Details

Gene
MTMR11
Gene Name
myotubularin related protein 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145862.1 2699 Missense Mutation CAC,CGC H637R NP_001139334.1
NM_181873.3 2699 Missense Mutation CAC,CGC H565R NP_870988.2
XM_006711137.1 2699 Missense Mutation CAC,CGC H565R XP_006711200.1
XM_011509099.2 2699 UTR 3 XP_011507401.1
Gene
SF3B4
Gene Name
splicing factor 3b subunit 4
There are no transcripts associated with this gene.

View Full Product Details