Product Details

SNP ID
rs145997850
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:89368526 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAATGTAACGTTATTTCTACTGG[A/G]AGGCTCTTCTAGGTTGGCAGTTGCC
Phenotype
MIM: 612467
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GBP6 PubMed Links

Gene Details

Gene
GBP6
Gene Name
guanylate binding protein family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320257.1 Intron NP_001307186.1
NM_198460.2 Intron NP_940862.2
XM_011540835.2 Intron XP_011539137.1

View Full Product Details