Product Details

SNP ID
rs146016415
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:111441265 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACTTGCAGGTCCAGGGGCTGGGA[A/G]AGGTTCTGTGGGCACAACGTGGTGG
Phenotype
MIM: 603270 MIM: 611734
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP5F1 PubMed Links

Gene Details

Gene
ATP5F1
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit B1
There are no transcripts associated with this gene.

Gene
WDR77
Gene Name
WD repeat domain 77
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317062.1 1169 Missense Mutation CTC,TTC L344F NP_001303991.1
NM_001317063.1 1169 Missense Mutation CTC,TTC L330F NP_001303992.1
NM_001317064.1 1169 Missense Mutation CTC,TTC L268F NP_001303993.1
NM_024102.3 1169 Missense Mutation CTC,TTC L332F NP_077007.1

View Full Product Details