Product Details

SNP ID
rs146068024
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:247531841 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGACCTGATCTTCAACACGGCC[C/T]GGGCAATGTGGCCGTAAGAGACCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GCSAML PubMed Links

Gene Details

Gene
GCSAML
Gene Name
germinal center associated signaling and motility like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281834.1 1033 Intron NP_001268763.1
NM_001281835.1 1033 Intron NP_001268764.1
NM_001281836.1 1033 Intron NP_001268765.1
NM_001281837.1 1033 Intron NP_001268766.1
NM_001281838.1 1033 Intron NP_001268767.1
NM_001281853.1 1033 Intron NP_001268782.1
NM_145278.4 1033 Intron NP_660321.1
XM_017000399.1 1033 Intron XP_016855888.1
Gene
GCSAML-AS1
Gene Name
GCSAML antisense RNA 1
There are no transcripts associated with this gene.

Gene
OR2C3
Gene Name
olfactory receptor family 2 subfamily C member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198074.4 1033 Missense Mutation CAG,CGG Q224R NP_932340.3

View Full Product Details