Product Details

SNP ID
rs146079560
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:229594363 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGGTTACAGGCCATGAACTGCAC[A/G]CTCAGGACGTTGCTCATCTGCCCGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TAF5L PubMed Links

Gene Details

Gene
TAF5L
Gene Name
TATA-box binding protein associated factor 5 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025247.1 1836 Intron NP_001020418.1
NM_014409.3 1836 Silent Mutation AGC,AGT S568S NP_055224.1
XM_005273099.4 1836 Silent Mutation AGC,AGT S568S XP_005273156.1
XM_005273100.2 1836 Silent Mutation AGC,AGT S476S XP_005273157.1
XM_011544162.1 1836 Silent Mutation AGC,AGT S497S XP_011542464.1
XM_011544163.1 1836 Silent Mutation AGC,AGT S476S XP_011542465.1
XM_011544164.2 1836 Silent Mutation AGC,AGT S476S XP_011542466.1
XM_017001045.1 1836 Silent Mutation AGC,AGT S476S XP_016856534.1

View Full Product Details