Product Details

SNP ID
rs146096083
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:219915560 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACTTCAATAGCCACTTCTCTTGC[A/G]TCTCTTCTACTGAGGCCATCACTTC
Phenotype
MIM: 611146
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC30A10 PubMed Links

Gene Details

Gene
SLC30A10
Gene Name
solute carrier family 30 member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018713.2 1894 Silent Mutation GAC,GAT D449D NP_061183.2
XM_006711437.3 1894 Silent Mutation GAC,GAT D386D XP_006711500.2
XM_017001684.1 1894 Silent Mutation GAC,GAT D290D XP_016857173.1

View Full Product Details