Product Details

SNP ID
rs146139739
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:8949318 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCACTACCCCGCCTGTGGGGGCCA[C/G]AGACAGTCGCCTATCAACCTACAGA
Phenotype
MIM: 114780
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CA6 PubMed Links

Gene Details

Gene
CA6
Gene Name
carbonic anhydrase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001215.3 236 Missense Mutation CAC,CAG H45Q NP_001206.2
NM_001270500.1 236 Missense Mutation CAC,CAG H45Q NP_001257429.1
NM_001270501.1 236 Intron NP_001257430.1
NM_001270502.1 236 Intron NP_001257431.1
XM_011542083.2 236 Missense Mutation CAC,CAG H49Q XP_011540385.1
XM_011542084.2 236 Missense Mutation CAC,CAG H49Q XP_011540386.1

View Full Product Details