Product Details

SNP ID
rs146203556
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:1387436 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACTTGTGTGGCTTCTGGGGGTAC[G/T]TGCAGTCCTTGGACTTCCGGGAGCC
Phenotype
MIM: 613482
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCNL2 PubMed Links

Gene Details

Gene
CCNL2
Gene Name
cyclin L2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039577.3 1274 Intron NP_001034666.1
NM_001320153.1 1274 Missense Mutation AAG,ACG K231T NP_001307082.1
NM_001320155.1 1274 Missense Mutation AAG,ACG K231T NP_001307084.1
NM_030937.4 1274 Missense Mutation AAG,ACG K453T NP_112199.2
XM_011542216.2 1274 Missense Mutation AAG,ACG K486T XP_011540518.1
XM_011542219.2 1274 Missense Mutation AAG,ACG K322T XP_011540521.1
XM_011542221.2 1274 Missense Mutation AAG,ACG K231T XP_011540523.1
XM_017002420.1 1274 Missense Mutation AAG,ACG K331T XP_016857909.1
XM_017002421.1 1274 Missense Mutation AAG,ACG K322T XP_016857910.1
XM_017002422.1 1274 Missense Mutation AAG,ACG K231T XP_016857911.1
XM_017002423.1 1274 Intron XP_016857912.1

View Full Product Details