Product Details

SNP ID
rs146236511
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:25246561 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAAGCCGTAGTAGCGCTGTCCCC[C/G]CGCGATCGCGTACGCCCTTCCGCAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RSRP1 PubMed Links

Gene Details

Gene
RSRP1
Gene Name
arginine and serine rich protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321772.1 645 Missense Mutation CGG,GGG R135G NP_001308701.1
NM_020317.4 645 Missense Mutation CGG,GGG R135G NP_064713.3
XM_011541797.1 645 Missense Mutation CGG,GGG R135G XP_011540099.1

View Full Product Details