Product Details

SNP ID
rs146293191
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:34865029 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGACTGGAGCAGAAGGAACATTGT[A/G]CCCCTAGGGGAAGGGAAACCAATTA
Phenotype
MIM: 611413
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DLGAP3 PubMed Links

Gene Details

Gene
DLGAP3
Gene Name
DLG associated protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080418.2 3972 Intron NP_001073887.1
XM_011541879.2 3972 UTR 3 XP_011540181.1
XM_011541880.2 3972 UTR 3 XP_011540182.1
Gene
SMIM12
Gene Name
small integral membrane protein 12
There are no transcripts associated with this gene.

View Full Product Details